Product Details

SNP ID
rs7352934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:57392678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGACCATGGCCGACCGGGCGGCA[C/G]CTGAGAGGGCTTGCAAAGACCCGAA
Phenotype
MIM: 612428
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100291105 PubMed Links

Gene Details

Gene
LOC100291105
Gene Name
uncharacterized LOC100291105
There are no transcripts associated with this gene.

Gene
RBM38
Gene Name
RNA binding motif protein 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291780.1 480 Missense Mutation CCT,GCT P120A NP_001278709.1
NM_017495.5 480 Missense Mutation CCT,GCT P88A NP_059965.2
NM_183425.2 480 Missense Mutation CCT,GCT P88A NP_906270.1
XM_005260446.3 480 Missense Mutation CCT,GCT P120A XP_005260503.1
XM_011528885.2 480 Missense Mutation CCT,GCT P120A XP_011527187.1

View Full Product Details