Product Details

SNP ID
rs6512729
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:51600017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATGCCGGCATGTAGTCCATCCT[A/G]GGAGGGGAGAAATCTTCACCACTGG
Phenotype
MIM: 609126
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP9A PubMed Links

Gene Details

Gene
ATP9A
Gene Name
ATPase phospholipid transporting 9A (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006045.2 4408 UTR 3 NP_006036.1
XM_011528481.2 4408 UTR 3 XP_011526783.1

View Full Product Details