Product Details

SNP ID
rs6094374
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46540206 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTATATATATGGTATCATGTTGTTT[A/C]AAACGCAACAAACAATATTACATAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OCSTAMP PubMed Links

Gene Details

Gene
OCSTAMP
Gene Name
osteoclast stimulatory transmembrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080721.2 Intron NP_542452.1
XM_011528532.2 Intron XP_011526834.1

View Full Product Details