Product Details

SNP ID
rs6426328
Assay Type
Functionally tested
NCBI dbSNP Submissions
38
Location
Chr.1:246727958 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAAGGGCTTCAGCTGATTTAAATT[G/T]GAAGTTCTTTGGCTTCTGAGTGGAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SCCPDH PubMed Links

Gene Details

Gene
SCCPDH
Gene Name
saccharopine dehydrogenase (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016002.2 Intron NP_057086.2

View Full Product Details