Product Details

SNP ID
rs8009058
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:80956524 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGATTAGAAACCTGATGTTAAATAA[C/G]ATTATACACACACAACCTGTTTCTT
Phenotype
MIM: 603372
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CEP128 PubMed Links

Gene Details

Gene
CEP128
Gene Name
centrosomal protein 128
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152446.3 Intron NP_689659.2
XM_006720056.3 Intron XP_006720119.1
XM_011536490.2 Intron XP_011534792.1
XM_011536491.2 Intron XP_011534793.1
XM_011536492.2 Intron XP_011534794.1
XM_011536493.2 Intron XP_011534795.1
XM_011536495.2 Intron XP_011534797.1
XM_017021042.1 Intron XP_016876531.1
XM_017021043.1 Intron XP_016876532.1
Gene
TSHR
Gene Name
thyroid stimulating hormone receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000369.2 Intron NP_000360.2
NM_001018036.2 Intron NP_001018046.1
NM_001142626.2 Intron NP_001136098.1
XM_005268037.4 Intron XP_005268094.1
XM_005268039.1 Intron XP_005268096.1
XM_006720245.1 Intron XP_006720308.1
XM_011537119.2 Intron XP_011535421.1

View Full Product Details