Product Details

SNP ID
rs7834318
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143555087 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGTGGCTGCAGCATTCCAACGCA[A/C]ACACCACCAGAAATCCCCTTCAGCA
Phenotype
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
GSDMD PubMed Links

Gene Details

Gene
GSDMD
Gene Name
gasdermin D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166237.1 403 UTR 5 NP_001159709.1
NM_024736.6 403 Intron NP_079012.3
XM_011517301.1 403 Intron XP_011515603.1
Gene
ZC3H3
Gene Name
zinc finger CCCH-type containing 3
There are no transcripts associated with this gene.

View Full Product Details