Product Details

SNP ID
rs8047946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67228452 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTGGGAGGTGGATGAGTGCGTA[A/G]GGTCAGTGGCTGGCCTTTGATACCC
Phenotype
MIM: 606881
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FHOD1 PubMed Links

Gene Details

Gene
FHOD1
Gene Name
formin homology 2 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318202.1 Intron NP_001305131.1
NM_013241.2 Intron NP_037373.2
XM_006721180.1 Intron XP_006721243.1
XM_011523043.2 Intron XP_011521345.1
XM_011523044.1 Intron XP_011521346.1
XM_011523045.2 Intron XP_011521347.1
Gene
LOC105369155
Gene Name
uncharacterized LOC105369155
There are no transcripts associated with this gene.

Gene
LRRC29
Gene Name
leucine rich repeat containing 29
There are no transcripts associated with this gene.

Gene
TMEM208
Gene Name
transmembrane protein 208
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318217.1 Intron NP_001305146.1
NM_014187.3 Intron NP_054906.2

View Full Product Details