Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001321708.1 | 6794 | Intron | NP_001308637.1 | ||
NM_001321709.1 | 6794 | Intron | NP_001308638.1 | ||
NM_001321710.1 | 6794 | Intron | NP_001308639.1 | ||
NM_004717.3 | 6794 | Intron | NP_004708.1 | ||
XM_011516682.2 | 6794 | Intron | XP_011514984.2 | ||
XM_011516684.2 | 6794 | UTR 3 | XP_011514986.2 | ||
XM_017012780.1 | 6794 | UTR 3 | XP_016868269.1 | ||
XM_017012781.1 | 6794 | UTR 3 | XP_016868270.1 | ||
XM_017012782.1 | 6794 | UTR 3 | XP_016868271.1 | ||
XM_017012783.1 | 6794 | UTR 3 | XP_016868272.1 | ||
XM_017012784.1 | 6794 | UTR 3 | XP_016868273.1 | ||
XM_017012785.1 | 6794 | UTR 3 | XP_016868274.1 | ||
XM_017012786.1 | 6794 | UTR 3 | XP_016868275.1 | ||
XM_017012787.1 | 6794 | UTR 3 | XP_016868276.1 | ||
XM_017012788.1 | 6794 | UTR 3 | XP_016868277.1 | ||
XM_017012789.1 | 6794 | UTR 3 | XP_016868278.1 |