Product Details
- SNP ID
-
rs7249755
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:10043346 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATATGGATTGATGTGAATGACAGA[C/T]GCATAGGGATGGTAAGCCAAACTCC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C3P1
PubMed Links
Gene Details
- Gene
- C3P1
- Gene Name
- complement component 3 precursor pseudogene
There are no transcripts associated with this gene.
- Gene
- MIR5589
- Gene Name
- microRNA 5589
There are no transcripts associated with this gene.
View Full Product Details