Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001258452.1 | 172 | Intron | NP_001245381.1 | ||
NM_001258453.1 | 172 | Intron | NP_001245382.1 | ||
NM_032807.4 | 172 | Intron | NP_116196.3 | ||
NM_178150.2 | 172 | Intron | NP_835363.1 | ||
XM_011519748.2 | 172 | UTR 5 | XP_011518050.1 | ||
XM_017016842.1 | 172 | UTR 5 | XP_016872331.1 | ||
XM_017016843.1 | 172 | UTR 5 | XP_016872332.1 | ||
XM_017016844.1 | 172 | Intron | XP_016872333.1 | ||
XM_017016845.1 | 172 | UTR 5 | XP_016872334.1 | ||
XM_017016846.1 | 172 | Intron | XP_016872335.1 |