Product Details

SNP ID
rs6478712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:125752329 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGGGAAGAGAAAGGCCTGCTTGG[C/T]TGGTTGGGCTGGTTGAAGTATTTTT
Phenotype
MIM: 176312
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC51145 PubMed Links

Gene Details

Gene
LOC51145
Gene Name
uncharacterized LOC51145
There are no transcripts associated with this gene.

Gene
PBX3
Gene Name
PBX homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134778.1 Intron NP_001128250.1
NM_006195.5 Intron NP_006186.1
XM_006717130.1 Intron XP_006717193.1
XM_006717131.1 Intron XP_006717194.1
XM_006717132.1 Intron XP_006717195.1
XM_006717133.1 Intron XP_006717196.1
XM_011518755.1 Intron XP_011517057.1
XM_017014786.1 Intron XP_016870275.1

View Full Product Details