Product Details

SNP ID
rs8081683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81714048 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCATCTGGCCGGGACGGGGGCTGCT[A/G]CCTGTGTGGCCCAGTCCCTCTGTTC
Phenotype
MIM: 602375 MIM: 606794
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL12 PubMed Links

Gene Details

Gene
MRPL12
Gene Name
mitochondrial ribosomal protein L12
There are no transcripts associated with this gene.

Gene
SLC25A10
Gene Name
solute carrier family 25 member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270888.1 Intron NP_001257817.1
NM_001270953.1 Intron NP_001257882.1
NM_012140.4 Intron NP_036272.2
XM_017024220.1 Intron XP_016879709.1

View Full Product Details