Product Details

SNP ID
rs6982970
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:126555893 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTTGGTCTGCTGCTCCCGGACCT[C/T]GTCATGGATTCACCAGCCCTGCGCT
Phenotype
MIM: 609483
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM84B PubMed Links

Gene Details

Gene
FAM84B
Gene Name
family with sequence similarity 84 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174911.4 2026 UTR 3 NP_777571.1
XM_017013107.1 2026 UTR 3 XP_016868596.1
XM_017013108.1 2026 UTR 3 XP_016868597.1

View Full Product Details