Product Details
- SNP ID
-
rs8065479
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:59336686 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCTAGCTATTAATAACAGCACTAA[C/T]ATTAATAGGTGGATTCCCCTCTCTC
- Phenotype
-
MIM: 609723
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
YPEL2
PubMed Links
Gene Details
- Gene
- YPEL2
- Gene Name
- yippee like 2
View Full Product Details