Product Details

SNP ID
rs7301041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10220725 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAAGGCTCTGAGAGATTAGATACC[A/T]CTTGTTTTTTAGGGATTAGCCACTC
Phenotype
MIM: 607420
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GABARAPL1 PubMed Links

Gene Details

Gene
GABARAPL1
Gene Name
GABA type A receptor associated protein like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031412.2 744 Intron NP_113600.1
XM_005253344.4 744 UTR 3 XP_005253401.1

View Full Product Details