Product Details

SNP ID
rs7966726
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110445416 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTCAGAAGATTGTTAGGCATTT[C/T]GTACCAAAATTTGAAAATAATGGGT
Phenotype
MIM: 604225
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARPC3 PubMed Links

Gene Details

Gene
ARPC3
Gene Name
actin related protein 2/3 complex subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278556.1 Intron NP_001265485.1
NM_001287222.1 Intron NP_001274151.1
Gene
GPN3
Gene Name
GPN-loop GTPase 3
There are no transcripts associated with this gene.

View Full Product Details