Product Details
- SNP ID
-
rs7962830
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:111456411 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGGTGGTAAGAGCAAGGCTGGAG[A/G]TGCCTCCCATCTGGACAACTCCTTT
- Phenotype
-
MIM: 601517
MIM: 605093
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ATXN2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112609183] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATXN2
- Gene Name
- ataxin 2
- Gene
- SH2B3
- Gene Name
- SH2B adaptor protein 3
There are no transcripts associated with this gene.
View Full Product Details