Product Details

SNP ID
rs7298769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:113231731 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCAAGACTCAGACATCTTTGGGCT[C/G]TAAGTGCGGCTGCTGCAGTGCTAAT
Phenotype
MIM: 609666
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TPCN1 PubMed Links

Gene Details

Gene
TPCN1
Gene Name
two pore segment channel 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143819.2 Intron NP_001137291.1
NM_001301214.1 Intron NP_001288143.1
NM_017901.5 Intron NP_060371.2
XM_005253906.4 Intron XP_005253963.1
XM_011538490.2 Intron XP_011536792.1
XM_011538492.2 Intron XP_011536794.1
XM_011538493.2 Intron XP_011536795.1
XM_017019480.1 Intron XP_016874969.1
XM_017019481.1 Intron XP_016874970.1
XM_017019482.1 Intron XP_016874971.1

View Full Product Details