Product Details

SNP ID
rs7952886
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:113302166 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAAATAGGGTCTTTGCAAATATAA[G/T]AAGCTAAGGGTTTCAAGATACAATC
Phenotype
MIM: 609841 MIM: 609666
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC8B1 PubMed Links

Gene Details

Gene
SLC8B1
Gene Name
solute carrier family 8 member B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006719607.2 Intron XP_006719670.1
XM_011538749.2 Intron XP_011537051.1
XM_011538750.2 Intron XP_011537052.1
XM_011538752.2 Intron XP_011537054.1
XM_017019978.1 Intron XP_016875467.1
Gene
TPCN1
Gene Name
two pore segment channel 1
There are no transcripts associated with this gene.

View Full Product Details