Product Details

SNP ID
rs8082472
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:21241205 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCCTGGCTCTCAAGGTCCATGCT[G/T]GCCCTCAGGGGCTTCCTGTGGGGGA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NATD1 PubMed Links

Gene Details

Gene
NATD1
Gene Name
N-acetyltransferase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152914.2 2452 UTR 3 NP_690878.2

View Full Product Details