Product Details
- SNP ID
-
rs7687648
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:184650119 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CGAGCTGCGTGGAGTTCCCAAGGAG[G/T]CAGTTCTCAGCACTTGCTGGTCACT
- Phenotype
-
MIM: 600636
MIM: 615421
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CASP3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79773844] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CASP3
- Gene Name
- caspase 3
- Gene
- PRIMPOL
- Gene Name
- primase and DNA directed polymerase
View Full Product Details