Product Details
- SNP ID
-
rs6554358
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:56506410 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTGTTGAGGGACAAGAGATCTCGG[A/T]CTTCATGGGGACATGGGGAACGGCC
- Phenotype
-
MIM: 612405
MIM: 602122
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ARL9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76513197] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARL9
- Gene Name
- ADP ribosylation factor like GTPase 9
- Gene
- SRP72
- Gene Name
- signal recognition particle 72
There are no transcripts associated with this gene.
View Full Product Details