Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001282480.1 | 2961 | UTR 3 | NP_001269409.1 | ||
NM_001282481.1 | 2961 | UTR 3 | NP_001269410.1 | ||
NM_001282482.1 | 2961 | UTR 3 | NP_001269411.1 | ||
NM_001282483.1 | 2961 | UTR 3 | NP_001269412.1 | ||
NM_001282485.1 | 2961 | UTR 3 | NP_001269414.1 | ||
NM_006259.2 | 2961 | UTR 3 | NP_006250.1 | ||
XM_005263126.3 | 2961 | UTR 3 | XP_005263183.1 | ||
XM_017008413.1 | 2961 | Intron | XP_016863902.1 | ||
XM_017008414.1 | 2961 | Intron | XP_016863903.1 | ||
XM_017008415.1 | 2961 | Intron | XP_016863904.1 | ||
XM_017008416.1 | 2961 | Intron | XP_016863905.1 | ||
XM_017008417.1 | 2961 | Intron | XP_016863906.1 | ||
XM_017008418.1 | 2961 | Intron | XP_016863907.1 |