Product Details

SNP ID
rs5746136
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:159682052 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAGTATCACTGCAGAGGATTACT[C/T]GACAAAAAGTGCAATTTTCTATAGA
Phenotype
MIM: 147460
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SOD2 PubMed Links

Gene Details

Gene
SOD2
Gene Name
superoxide dismutase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000636.3 1091 UTR 3 NP_000627.2
NM_001024465.2 1091 Intron NP_001019636.1
NM_001024466.2 1091 Intron NP_001019637.1
NM_001322814.1 1091 UTR 3 NP_001309743.1
NM_001322815.1 1091 UTR 3 NP_001309744.1
NM_001322816.1 1091 Intron NP_001309745.1
NM_001322817.1 1091 Intron NP_001309746.1
NM_001322819.1 1091 UTR 3 NP_001309748.1
NM_001322820.1 1091 UTR 3 NP_001309749.1

View Full Product Details