Product Details

SNP ID
rs7757796
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:108853493 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTATGGACTTTGAGTTTTTTTTTA[A/C]GAAACACCTAATGCTATATACTCAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARMC2 PubMed Links

Gene Details

Gene
ARMC2
Gene Name
armadillo repeat containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286609.1 Intron NP_001273538.1
NM_032131.5 Intron NP_115507.4
XM_005267154.3 Intron XP_005267211.1
XM_005267155.2 Intron XP_005267212.1
XM_005267157.3 Intron XP_005267214.1
XM_006715573.2 Intron XP_006715636.1
XM_006715574.2 Intron XP_006715637.1
XM_011536166.1 Intron XP_011534468.1
XM_011536167.1 Intron XP_011534469.1
XM_011536168.2 Intron XP_011534470.1
XM_011536170.2 Intron XP_011534472.1
XM_011536172.2 Intron XP_011534474.1
XM_011536176.2 Intron XP_011534478.1
XM_017011350.1 Intron XP_016866839.1
XM_017011351.1 Intron XP_016866840.1
XM_017011352.1 Intron XP_016866841.1

View Full Product Details