Product Details

SNP ID
rs8127692
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:42373273 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCCTGCTGGAAAGCCTCCCAGGA[A/G]AGGAAGTCTCTTCTGGGTCTCCCCT
Phenotype
MIM: 113710 MIM: 605511
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TFF1 PubMed Links

Gene Details

Gene
TFF1
Gene Name
trefoil factor 1
There are no transcripts associated with this gene.

Gene
TMPRSS3
Gene Name
transmembrane protease, serine 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256317.1 Intron NP_001243246.1
NM_024022.2 Intron NP_076927.1
NM_032404.2 Intron NP_115780.1
NM_032405.1 Intron NP_115781.1

View Full Product Details