Product Details

SNP ID
rs7281126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:40385254 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCTTAAGATCTCTCTATGTTGA[G/T]ATAAAGAAATCTAGCTCATTCTTTT
Phenotype
MIM: 602523
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DSCAM PubMed Links

Gene Details

Gene
DSCAM
Gene Name
DS cell adhesion molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271534.1 Intron NP_001258463.1
NM_001389.3 Intron NP_001380.2
XM_017028281.1 Intron XP_016883770.1
Gene
DSCAM-AS1
Gene Name
DSCAM antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details