Product Details
- SNP ID
-
rs6736360
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:189884023 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAACATTTACACCCAGATGCTTAAT[A/C]AGACAAAGTAATGACTGGTAATTTA
- Phenotype
-
MIM: 615117
MIM: 600258
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C2orf88
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs138997788] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C2orf88
- Gene Name
- chromosome 2 open reading frame 88
- Gene
- PMS1
- Gene Name
- PMS1 homolog 1, mismatch repair system component
There are no transcripts associated with this gene.
View Full Product Details