Product Details

SNP ID
rs7461388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:380328 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCCAGCTGATGGAACGTAAACAG[A/C]AACCAGCACATCAGCTCCAAGTGGA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM87A PubMed Links

Gene Details

Gene
FAM87A
Gene Name
family with sequence similarity 87 member A
There are no transcripts associated with this gene.

View Full Product Details