Product Details

SNP ID
rs3739579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:8314988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGTACAAATCACTTTTTATATA[G/T]TTTGATTTTTTTTACCATTGTAACT
Phenotype
MIM: 601598
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PTPRD PubMed Links

Gene Details

Gene
PTPRD
Gene Name
protein tyrosine phosphatase, receptor type D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040712.2 9724 UTR 3 NP_001035802.1
NM_001171025.1 9724 UTR 3 NP_001164496.1
NM_002839.3 9724 UTR 3 NP_002830.1
NM_130391.3 9724 UTR 3 NP_569075.2
NM_130392.3 9724 UTR 3 NP_569076.2
NM_130393.3 9724 UTR 3 NP_569077.2
XM_006716817.3 9724 UTR 3 XP_006716880.1
XM_006716823.2 9724 UTR 3 XP_006716886.1
XM_006716825.3 9724 UTR 3 XP_006716888.1
XM_006716827.3 9724 UTR 3 XP_006716890.1
XM_006716832.3 9724 UTR 3 XP_006716895.1
XM_006716833.3 9724 UTR 3 XP_006716896.1
XM_006716834.3 9724 UTR 3 XP_006716897.1
XM_006716835.3 9724 UTR 3 XP_006716898.1
XM_006716837.3 9724 UTR 3 XP_006716900.1
XM_006716838.3 9724 UTR 3 XP_006716901.1
XM_006716839.3 9724 UTR 3 XP_006716902.1
XM_011517992.2 9724 UTR 3 XP_011516294.1
XM_017014958.1 9724 UTR 3 XP_016870447.1
XM_017014959.1 9724 UTR 3 XP_016870448.1
XM_017014960.1 9724 UTR 3 XP_016870449.1
XM_017014961.1 9724 UTR 3 XP_016870450.1
XM_017014962.1 9724 UTR 3 XP_016870451.1
XM_017014963.1 9724 UTR 3 XP_016870452.1
XM_017014964.1 9724 UTR 3 XP_016870453.1
XM_017014965.1 9724 UTR 3 XP_016870454.1
XM_017014966.1 9724 UTR 3 XP_016870455.1
XM_017014967.1 9724 UTR 3 XP_016870456.1
XM_017014968.1 9724 UTR 3 XP_016870457.1
XM_017014969.1 9724 UTR 3 XP_016870458.1
XM_017014970.1 9724 UTR 3 XP_016870459.1
XM_017014971.1 9724 UTR 3 XP_016870460.1
XM_017014972.1 9724 UTR 3 XP_016870461.1
XM_017014973.1 9724 UTR 3 XP_016870462.1
XM_017014974.1 9724 UTR 3 XP_016870463.1
XM_017014975.1 9724 UTR 3 XP_016870464.1
XM_017014976.1 9724 UTR 3 XP_016870465.1
XM_017014977.1 9724 UTR 3 XP_016870466.1
XM_017014978.1 9724 Intron XP_016870467.1
XM_017014979.1 9724 UTR 3 XP_016870468.1
XM_017014980.1 9724 UTR 3 XP_016870469.1
XM_017014981.1 9724 UTR 3 XP_016870470.1
XM_017014982.1 9724 UTR 3 XP_016870471.1
XM_017014983.1 9724 UTR 3 XP_016870472.1
XM_017014984.1 9724 UTR 3 XP_016870473.1
XM_017014985.1 9724 UTR 3 XP_016870474.1
XM_017014986.1 9724 UTR 3 XP_016870475.1
XM_017014987.1 9724 UTR 3 XP_016870476.1
XM_017014988.1 9724 UTR 3 XP_016870477.1
XM_017014989.1 9724 UTR 3 XP_016870478.1
XM_017014990.1 9724 Intron XP_016870479.1
XM_017014991.1 9724 UTR 3 XP_016870480.1
XM_017014992.1 9724 Intron XP_016870481.1
XM_017014993.1 9724 UTR 3 XP_016870482.1
XM_017014994.1 9724 UTR 3 XP_016870483.1
XM_017014995.1 9724 UTR 3 XP_016870484.1

View Full Product Details