Product Details

SNP ID
rs7022122
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:2624186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTTTCACTTTGTCACATGTGAA[A/G]TATTTGACATGCCATGAACAATGGA
Phenotype
MIM: 192977
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VLDLR PubMed Links

Gene Details

Gene
VLDLR
Gene Name
very low density lipoprotein receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018056.2 Intron NP_001018066.1
NM_001322225.1 Intron NP_001309154.1
NM_001322226.1 Intron NP_001309155.1
NM_003383.4 Intron NP_003374.3
Gene
VLDLR-AS1
Gene Name
VLDLR antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details