Product Details
- SNP ID
-
rs6499512
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:71369332 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAGGTGGTGTCCTCTAATCTGGGTA[C/T]GACAGAGGGAGGAGAAGAACCAGGT
- Phenotype
-
MIM: 114051
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CALB2
PubMed Links
Gene Details
- Gene
- CALB2
- Gene Name
- calbindin 2
View Full Product Details