Product Details

SNP ID
rs8136547
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:27978590 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGAGCAGGATGAGGACAGCGTGG[A/G]TGACACCTTTGTCTCCTGCTAAAAC
Phenotype
MIM: 615098
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTC28 PubMed Links

Gene Details

Gene
TTC28
Gene Name
tetratricopeptide repeat domain 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145418.1 11219 UTR 3 NP_001138890.1
XM_005261405.2 11219 UTR 3 XP_005261462.1
XM_006724171.3 11219 UTR 3 XP_006724234.1
XM_011530018.2 11219 UTR 3 XP_011528320.1
XM_011530019.2 11219 Intron XP_011528321.1
XM_011530020.1 11219 Intron XP_011528322.1
XM_011530021.2 11219 Intron XP_011528323.1
XM_011530022.1 11219 Intron XP_011528324.1
XM_017028673.1 11219 UTR 3 XP_016884162.1
Gene
TTC28-AS1
Gene Name
TTC28 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details