Product Details
- SNP ID
-
rs7082689
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:123157192 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTTGAACCACAGTGGAACCAAGTG[C/G]AGTAATGAGAGGACTTCACTCCCAG
- Phenotype
-
MIM: 603719
MIM: 600647
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
BUB3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75868905] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BUB3
- Gene Name
- BUB3, mitotic checkpoint protein
- Gene
- HMX2
- Gene Name
- H6 family homeobox 2
There are no transcripts associated with this gene.
View Full Product Details