Product Details

SNP ID
rs7905088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:123147184 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCTAAAACCGAGTCCTGTCACCA[C/G]TTCCTTATCTTCTTCCTGGGAGTAA
Phenotype
MIM: 603719 MIM: 600647 MIM: 613380
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BUB3 PubMed Links

Gene Details

Gene
BUB3
Gene Name
BUB3, mitotic checkpoint protein
There are no transcripts associated with this gene.

Gene
HMX2
Gene Name
H6 family homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005519.1 Intron NP_005510.1
XM_005269743.3 Intron XP_005269800.1
Gene
HMX3
Gene Name
H6 family homeobox 3
There are no transcripts associated with this gene.

View Full Product Details