Product Details
- SNP ID
-
rs7915885
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:100469594 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCTCAAATTGTACAGTGAATAGTT[C/T]CAAGTTCAATCCCCTCAGGTATTAT
- Phenotype
-
MIM: 601396
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WNT8B
PubMed Links
Gene Details
- Gene
- WNT8B
- Gene Name
- Wnt family member 8B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003393.3 |
|
Intron |
|
|
NP_003384.2 |
View Full Product Details