Product Details

SNP ID
rs6596167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:134161558 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACGCTCAACTTTACTAGAAGCAAA[A/G]CTAACACTACAGATACTTCTCTAGA
Phenotype
MIM: 601434
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SKP1 PubMed Links

Gene Details

Gene
SKP1
Gene Name
S-phase kinase-associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006930.3 Intron NP_008861.2
NM_170679.2 Intron NP_733779.1

View Full Product Details