Product Details

SNP ID
rs6557829
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22116457 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCGGGGGGACATGGACAGTGAGG[A/C]TCAAGATCACACATCCTCTCCCTGT
Phenotype
MIM: 602302 MIM: 615791
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HR PubMed Links

Gene Details

Gene
HR
Gene Name
hair growth associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005144.4 Intron NP_005135.2
NM_018411.4 Intron NP_060881.2
XM_005273569.1 Intron XP_005273626.1
XM_006716367.1 Intron XP_006716430.1
Gene
NUDT18
Gene Name
nudix hydrolase 18
There are no transcripts associated with this gene.

View Full Product Details