Product Details

SNP ID
rs7005304
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22057154 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGAAGAGGTAGGTGGCAGGGTCTC[A/C]GCCTCTCCTCCCCTCTGTGGGCACT
Phenotype
MIM: 125305 MIM: 603725
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DMTN PubMed Links

Gene Details

Gene
DMTN
Gene Name
dematin actin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114135.4 Intron NP_001107607.1
NM_001114136.2 Intron NP_001107608.1
NM_001114137.3 Intron NP_001107609.1
NM_001114138.2 Intron NP_001107610.1
NM_001114139.3 Intron NP_001107611.1
NM_001302816.2 Intron NP_001289745.1
NM_001302817.2 Intron NP_001289746.1
NM_001323378.1 Intron NP_001310307.1
NM_001323380.1 Intron NP_001310309.1
NM_001323381.1 Intron NP_001310310.1
NM_001323382.1 Intron NP_001310311.1
NM_001323383.1 Intron NP_001310312.1
NM_001323384.1 Intron NP_001310313.1
NM_001323385.1 Intron NP_001310314.1
NM_001323387.1 Intron NP_001310316.1
NM_001323388.1 Intron NP_001310317.1
NM_001323389.1 Intron NP_001310318.1
NM_001323390.1 Intron NP_001310319.1
NM_001323391.1 Intron NP_001310320.1
NM_001323392.1 Intron NP_001310321.1
NM_001323393.1 Intron NP_001310322.1
NM_001323394.1 Intron NP_001310323.1
NM_001323395.1 Intron NP_001310324.1
NM_001323396.1 Intron NP_001310325.1
NM_001323397.1 Intron NP_001310326.1
NM_001323398.1 Intron NP_001310327.1
NM_001323399.1 Intron NP_001310328.1
NM_001323400.1 Intron NP_001310329.1
NM_001323401.1 Intron NP_001310330.1
NM_001978.4 Intron NP_001969.2
XM_005273432.1 Intron XP_005273489.1
XM_005273433.1 Intron XP_005273490.1
XM_005273442.1 Intron XP_005273499.1
XM_011544435.1 Intron XP_011542737.1
XM_017013189.1 Intron XP_016868678.1
XM_017013190.1 Intron XP_016868679.1
XM_017013191.1 Intron XP_016868680.1
XM_017013192.1 Intron XP_016868681.1
XM_017013193.1 Intron XP_016868682.1
XM_017013194.1 Intron XP_016868683.1
XM_017013195.1 Intron XP_016868684.1
XM_017013196.1 Intron XP_016868685.1
XM_017013197.1 Intron XP_016868686.1
XM_017013198.1 Intron XP_016868687.1
Gene
FGF17
Gene Name
fibroblast growth factor 17
There are no transcripts associated with this gene.

View Full Product Details