Product Details

SNP ID
rs6805138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:45030046 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTTAGGGCAGCCTGGGTTTCCTC[A/G]TGGTGAAATGCTGGTGGTACAGAAT
Phenotype
MIM: 187520
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLEC3B PubMed Links

Gene Details

Gene
CLEC3B
Gene Name
C-type lectin domain family 3 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308394.1 Intron NP_001295323.1
NM_003278.2 Intron NP_003269.2
XM_017007116.1 Intron XP_016862605.1
XM_017007117.1 Intron XP_016862606.1

View Full Product Details