Product Details

SNP ID
rs7157920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24440230 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAGACAGCTTGCACCACAGCGCT[A/G]GGGAGAGGGATGAAGGCTCGGCGGC
Phenotype
MIM: 616162
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KHNYN PubMed Links

Gene Details

Gene
KHNYN
Gene Name
KH and NYN domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290256.1 727 UTR 3 NP_001277185.1
NM_001290257.1 727 UTR 3 NP_001277186.1
NM_015299.2 727 UTR 3 NP_056114.1
XM_005267474.4 727 UTR 3 XP_005267531.1
XM_011536590.2 727 UTR 3 XP_011534892.1
XM_017021114.1 727 Intron XP_016876603.1
Gene
LOC101927045
Gene Name
uncharacterized LOC101927045
There are no transcripts associated with this gene.

Gene
SDR39U1
Gene Name
short chain dehydrogenase/reductase family 39U member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290292.1 727 Silent Mutation CCC,CCT P163P NP_001277221.1
NM_001290293.2 727 Silent Mutation CCC,CCT P137P NP_001277222.1
NM_001308075.1 727 Silent Mutation CCC,CCT P163P NP_001295004.1
NM_020195.2 727 Silent Mutation CCC,CCT P245P NP_064580.2
XM_011536975.1 727 Silent Mutation CCC,CCT P268P XP_011535277.1
XM_011536976.1 727 Silent Mutation CCC,CCT P203P XP_011535278.1

View Full Product Details