Product Details

SNP ID
rs7159891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24440194 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGGCCCTCCAGCAGCATGATGGC[A/C]CGCTGTCGCCCAAAGACAGCTTGCA
Phenotype
MIM: 616162
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KHNYN PubMed Links

Gene Details

Gene
KHNYN
Gene Name
KH and NYN domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290256.1 763 UTR 3 NP_001277185.1
NM_001290257.1 763 UTR 3 NP_001277186.1
NM_015299.2 763 UTR 3 NP_056114.1
XM_005267474.4 763 UTR 3 XP_005267531.1
XM_011536590.2 763 UTR 3 XP_011534892.1
XM_017021114.1 763 Intron XP_016876603.1
Gene
LOC101927045
Gene Name
uncharacterized LOC101927045
There are no transcripts associated with this gene.

Gene
SDR39U1
Gene Name
short chain dehydrogenase/reductase family 39U member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290292.1 763 Silent Mutation CGG,CGT R175R NP_001277221.1
NM_001290293.2 763 Silent Mutation CGG,CGT R149R NP_001277222.1
NM_001308075.1 763 Silent Mutation CGG,CGT R175R NP_001295004.1
NM_020195.2 763 Silent Mutation CGG,CGT R257R NP_064580.2
XM_011536975.1 763 Silent Mutation CGG,CGT R280R XP_011535277.1
XM_011536976.1 763 Silent Mutation CGG,CGT R215R XP_011535278.1

View Full Product Details