Product Details

SNP ID
rs7945178
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124637997 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTCCATCTCCCACCCTCAAATAT[C/T]GAAGACTCCTGAATAATCTTTCATC
Phenotype
MIM: 610079 MIM: 610614
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SIAE PubMed Links

Gene Details

Gene
SIAE
Gene Name
sialic acid acetylesterase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199922.1 Intron NP_001186851.1
NM_170601.4 Intron NP_733746.1
XM_017017930.1 Intron XP_016873419.1
Gene
TBRG1
Gene Name
transforming growth factor beta regulator 1
There are no transcripts associated with this gene.

View Full Product Details