Product Details

SNP ID
rs7103953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:33096728 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGATACACTTAGACTGCCTTTAC[A/G]AAATGGAGCAATAAAAGCTTAATGA
Phenotype
MIM: 600367
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSTF3 PubMed Links

Gene Details

Gene
CSTF3
Gene Name
cleavage stimulation factor subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033505.1 Intron NP_001028677.1
NM_001033506.1 Intron NP_001028678.1
NM_001326.2 Intron NP_001317.1

View Full Product Details