Product Details

SNP ID
rs6659240
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:236404257 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATTTGATTTTACACTTAATAGTAT[C/G]GTAAACATGCAACTCCAAGTTCCTA
Phenotype
MIM: 606603
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EDARADD PubMed Links

Gene Details

Gene
EDARADD
Gene Name
EDAR associated death domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080738.3 Intron NP_542776.1
NM_145861.2 Intron NP_665860.2

View Full Product Details