Product Details

SNP ID
rs6720823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:235496752 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGATGTGGTTGCATTTTCCTTT[C/T]TCCCTTAAAAGAAAAAAAAAATGCG
Phenotype
MIM: 608651
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGAP1 PubMed Links

Gene Details

Gene
AGAP1
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037131.2 Intron NP_001032208.1
NM_001244888.1 Intron NP_001231817.1
NM_014914.4 Intron NP_055729.2
XM_005246059.4 Intron XP_005246116.1
XM_006712234.3 Intron XP_006712297.1
XM_006712235.3 Intron XP_006712298.1
XM_006712237.3 Intron XP_006712300.2
XM_006712239.3 Intron XP_006712302.2
XM_011510547.2 Intron XP_011508849.1
XM_011510548.2 Intron XP_011508850.1
XM_011510549.2 Intron XP_011508851.1
XM_017003282.1 Intron XP_016858771.1
Gene
AGAP1-IT1
Gene Name
AGAP1 intronic transcript 1
There are no transcripts associated with this gene.

View Full Product Details