Product Details

SNP ID
rs3986419
Assay Type
Functionally tested
NCBI dbSNP Submissions
44
Location
Chr.1:6561424 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTGACGCTACTGCTAAAGAGTCT[A/G]TCTGGGGCCGGGCACGGTAGCTCAT
Phenotype
MIM: 606225
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NOL9 PubMed Links

Gene Details

Gene
NOL9
Gene Name
nucleolar protein 9
There are no transcripts associated with this gene.

Gene
TAS1R1
Gene Name
taste 1 receptor member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138697.3 Intron NP_619642.2
NM_177540.2 Intron NP_803884.1
XM_011542203.2 Intron XP_011540505.1
XM_011542206.2 Intron XP_011540508.1
XM_017002402.1 Intron XP_016857891.1
XM_017002403.1 Intron XP_016857892.1

View Full Product Details