Product Details

SNP ID
rs6950085
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:76054230 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGTTGGAGTCTTTCGCAACGTAC[A/G]TAACCCCTTCCACCTCTTGTTAGGC
Phenotype
MIM: 154100 MIM: 616695
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MDH2 PubMed Links

Gene Details

Gene
MDH2
Gene Name
malate dehydrogenase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282403.1 Intron NP_001269332.1
NM_001282404.1 Intron NP_001269333.1
NM_005918.3 Intron NP_005909.2
Gene
STYXL1
Gene Name
serine/threonine/tyrosine interacting-like 1
There are no transcripts associated with this gene.

View Full Product Details