Product Details

SNP ID
rs74315454
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:42693169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCGGCCAGGCTGCGGATGGAACA[T/C]CACTTCTTGAAGACCCGCGTGAGCA
Phenotype
MIM: 607922
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
A4GALT PubMed Links

Gene Details

Gene
A4GALT
Gene Name
alpha 1,4-galactosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318038.1 1271 Missense Mutation NP_001304967.1
NM_017436.5 1271 Missense Mutation NP_059132.1
XM_005261644.2 1271 Missense Mutation XP_005261701.1
XM_005261646.4 1271 Missense Mutation XP_005261703.1
XM_005261647.2 1271 Missense Mutation XP_005261704.1
XM_005261648.3 1271 Missense Mutation XP_005261705.1
XM_006724265.3 1271 Nonsense Mutation XP_006724328.1
XM_006724266.3 1271 Missense Mutation XP_006724329.1
XM_011530233.2 1271 Missense Mutation XP_011528535.1
XM_017028831.1 1271 Missense Mutation XP_016884320.1

View Full Product Details