Product Details

SNP ID
rs7479108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60456046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGTTCTTGATCTTAATTCTAGAG[G/T]TTCCCTAGTCAGACTTCCTAATAGT
Phenotype
MIM: 112210 MIM: 606499
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MS4A1 PubMed Links

Gene Details

Gene
MS4A1
Gene Name
membrane spanning 4-domains A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021950.3 Intron NP_068769.2
NM_152866.2 Intron NP_690605.1
Gene
MS4A5
Gene Name
membrane spanning 4-domains A5
There are no transcripts associated with this gene.

View Full Product Details